2-Bromo-5-chloro-4-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC7174-1G PC7174-25G PC7174-5G
APOSPC7174-1GEA 0 EUR
APOSPC7174-1G APOSPC7174-25G APOSPC7174-5G
2-Bromo-5-chloro-4-fluoroaniline
2-Bromo-5-chloro-4-fluoroaniline
Formel: C₆H₄BrClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD10698796
CAS-Nummer: 85462-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR