3-(Trifluormethyl)-5-pyridinboronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-(Trifluoromethyl)pyridin-3-yl)boronic acid, (5-Trifluoromethylpyridin-3-yl)boronic acid

PC7177-1G
APOSPC7177-1GEA 121 EUR
APOSPC7177-1G
3-(Trifluormethyl)-5-pyridinboronsäure 95%
3-(Trifluormethyl)-5-pyridinboronsäure
Formel: C₆H₅BF₃NO₂
Molecular Weight: 190.9156 g/mol
Schmelzpunkt: 260…262.6 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09952041
CAS-Nummer: 947533-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR