2-Fluor-3-(trifluormethyl)phenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-3-(trifluoromethyl)benzeneboronic acid
PC7181-1G
PC7181-5G
PC7181-10G
APOSPC7181-1GEA
21.9
EUR
APOSPC7181-1G
APOSPC7181-5G
APOSPC7181-10G
2-Fluor-3-(trifluormethyl)phenylboronsäure 97%
2-Fluor-3-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ Schmelzpunkt: 60…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07784336 CAS-Nummer: 157834-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...