2-Brom-6-fluorphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Bromo-6-fluorophenyl)boronic acid, 2-Bromo-6-fluorobenzeneboronic acid
PC7182-100G
PC7182-25G
PC7182-5G
APOSPC7182-100GEA
210
EUR
APOSPC7182-100G
APOSPC7182-25G
APOSPC7182-5G
2-Brom-6-fluorphenylboronsäure 98%
2-Brom-6-fluorphenylboronsäure
Formel:
C₆H₅BBrFO₂ Schmelzpunkt: 138…145 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05664309 CAS-Nummer: 913835-80-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...