4-Hydrazino-8-(trifluoromethyl)quinoline

Lieferant: Apollo Scientific

PC7188-250MG
APOSPC7188-250MGEA 415 EUR
APOSPC7188-250MG
4-Hydrazino-8-(trifluoromethyl)quinoline
4-Hydrazino-8-(trifluoromethyl)quinoline
Formel: C₁₀H₈F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00272472
CAS-Nummer: 57369-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR