4-Amino-2-methyl-7-(trifluormethyl)chinolin
Lieferant: Apollo Scientific
|
Achtung
|
PC7199-250MG
APOSPC7199-250MGEA
145
EUR
APOSPC7199-250MG
4-Amino-2-methyl-7-(trifluormethyl)chinolin
4-Amino-2-methyl-7-(trifluoromethyl)quinoline
Formel:
C₁₁H₉F₃N₂ Schmelzpunkt: 158…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01870545 CAS-Nummer: 917562-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...