5-Brom-2-chlor-3-fluorpyridin 98%

Lieferant: Apollo Scientific
Achtung

PC7206-5G
APOSPC7206-5GEA 116 EUR
APOSPC7206-5G
5-Brom-2-chlor-3-fluorpyridin 98%
5-Brom-2-chlor-3-fluorpyridin
Formel: C₅H₂BrClFN
Molecular Weight: 210.43 g/mol
Schmelzpunkt: 50…52 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03411574
CAS-Nummer: 831203-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR