6-Chloro-1,3-dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC7212-1G
APOSPC7212-1GEA
538
EUR
APOSPC7212-1G
6-Chloro-1,3-dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine
6-Chloro-1,3-dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine
Formel:
C₉H₇ClF₃N₃ Schmelzpunkt: 49…51 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09817432 CAS-Nummer: 952183-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...