6,8-Difluor-4-chinolinol

Lieferant: Apollo Scientific
Achtung

PC7213-1G
APOSPC7213-1GEA 0 EUR
APOSPC7213-1G
6,8-Difluor-4-chinolinol
6,8-Difluor-4-chinolinol
Formel: C₉H₅F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00278786
CAS-Nummer: 243448-16-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR