4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde

Lieferant: Apollo Scientific

PC7215-1G
APOSPC7215-1GEA 270 EUR
APOSPC7215-1G
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
Formel: C₁₃H₇ClF₃NO
Molecular Weight: 285,66 g/mol
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607889
CAS-Nummer: 952182-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR