4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarbaldehyde

Lieferant: Apollo Scientific
Achtung

PC7216-1G
APOSPC7216-1GEA 328 EUR
APOSPC7216-1G
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarbaldehyde
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarbaldehyde
Formel: C₁₃H₇ClF₃NO₂
Molecular Weight: 301,65 g/mol
Siedepunkt: 106 °C/0,075mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD09607890
CAS-Nummer: 950994-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR