{1-[3-(Trifluoromethyl)benzyl]-1H-1,2,3-triazol-4-yl}methanol

Lieferant: Apollo Scientific

Synonyme: 4-(Hydroxymethyl)-1-[3-(trifluoromethyl)benzyl]-1H-1,2,3-triazole

PC7224-1G
APOSPC7224-1GEA 336 EUR
APOSPC7224-1G
{1-[3-(Trifluoromethyl)benzyl]-1H-1,2,3-triazol-4-yl}methanol
{1-[3-(Trifluoromethyl)benzyl]-1H-1,2,3-triazol-4-yl}methanol
Formel: C₁₁H₁₀F₃N₃O
Molecular Weight: 257,22 g/mol
Schmelzpunkt: 73…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607910
CAS-Nummer: 952183-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR