1-[3-(Trifluoromethyl)benzyl]-1H-1,2,3-triazole-4-carbaldehyde

Lieferant: Apollo Scientific

PC7225-1G
APOSPC7225-1GEA 538 EUR
APOSPC7225-1G
1-[3-(Trifluoromethyl)benzyl]-1H-1,2,3-triazole-4-carbaldehyde
1-[3-(Trifluoromethyl)benzyl]-1H-1,2,3-triazole-4-carbaldehyde
Formel: C₁₁H₈F₃N₃O
Molecular Weight: 255,2 g/mol
Schmelzpunkt: 94…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607914
CAS-Nummer: 952183-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR