4-[4-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde

Lieferant: Apollo Scientific

PC7226-1G
APOSPC7226-1GEA 270 EUR
APOSPC7226-1G
4-[4-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
4-[4-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
Formel: C₁₃H₈F₃NO
Schmelzpunkt: 66…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607891
CAS-Nummer: 952182-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR