4,6-Difluoro-1H-benzimidazole
Lieferant: Apollo Scientific
|
Achtung
|
PC7239-250MG
APOSPC7239-250MGEA
0
EUR
APOSPC7239-250MG
4,6-Difluoro-1H-benzimidazole
4,6-Difluoro-1H-benzimidazole
Formel:
C₇H₄F₂N₂ Schmelzpunkt: 235…238 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425145 CAS-Nummer: 2208-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...