4-(2-(Trifluormethyl)phenylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

PC7248-1G
APOSPC7248-1GEA 0 EUR
APOSPC7248-1G
4-(2-(Trifluormethyl)phenylcarbamoyl)phenylboronsäure 98%
4-(2-(Trifluormethyl)phenylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₁BF₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09027216
CAS-Nummer: 913835-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR