[2-(Trifluoromethoxy)phenyl]methanesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

PC7251-1G PC7251-250MG
APOSPC7251-1GEA 261 EUR
APOSPC7251-1G APOSPC7251-250MG
[2-(Trifluoromethoxy)phenyl]methanesulphonyl chloride
[2-(Trifluoromethoxy)phenyl]methanesulphonyl chloride
Formel: C₈H₆ClF₃O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD09749658
CAS-Nummer: 116827-38-4
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR