Perfluoro-3,6-dioxadecanoic acid

Lieferant: Apollo Scientific

PC7256-100G PC7256-25G PC7256-1G PC7256-5G
APOSPC7256-100GEA 0 EUR
APOSPC7256-100G APOSPC7256-25G APOSPC7256-1G APOSPC7256-5G
Perfluoro-3,6-dioxadecanoic acid
Perfluoro-3,6-dioxadecanoic acid
Formel: C₈HF₁₅O₄
Siedepunkt: 55 °C/6mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03094421
CAS-Nummer: 137780-69-9
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR