2-(Difluormethoxy)iodbenzol

Lieferant: Apollo Scientific
Achtung

PC7263-25G PC7263-5G
APOSPC7263-25GEA 0 EUR
APOSPC7263-25G APOSPC7263-5G
2-(Difluormethoxy)iodbenzol
2-(Difluormethoxy)iodbenzol
Formel: C₇H₅F₂IO
Lagertemperatur: Kühlschrank
MDL: MFCD09749662
CAS-Nummer: 920981-12-0
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR