6-Bromo-2-fluoro-3-methoxybenzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-Fluoro-3-methoxy-6-bromophenylboronic acid

PC7266-1G
APOSPC7266-1GEA 241 EUR
APOSPC7266-1G
6-Bromo-2-fluoro-3-methoxybenzeneboronic acid 98%
6-Bromo-2-fluoro-3-methoxybenzeneboronic acid
Formel: C₇H₇BBrFO₃
Molecular Weight: 248,842 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258756
CAS-Nummer: 871126-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR