2H-Tridecafluoro-2-methylpentane 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC7271-5G
APOSPC7271-5GEA
290
EUR
APOSPC7271-5G
2H-Tridecafluoro-2-methylpentane 97%
2H-Tridecafluoro-2-methylpentane
Formel:
C₆HF₁₃ Siedepunkt: 60…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09763637 CAS-Nummer: 30320-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...