4-Benzyloxy-2,3-difluorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(Benzyloxy)-2,3-difluorophenyl)boronic acid, 4-(Benzyloxy)-2,3-difluorophenylboronic acid

PC7274-1G
APOSPC7274-1GEA 29 EUR
APOSPC7274-1G
4-Benzyloxy-2,3-difluorphenylboronsäure 98%
4-Benzyloxy-2,3-difluorphenylboronsäure
Formel: C₁₃H₁₁BF₂O₃
Molecular Weight: 264,0325 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07644469
CAS-Nummer: 156635-87-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR