2,3-Difluor-4-ethoxyphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Difluoro-4-ethoxybenzeneboronic acid, 4-Ethoxy-2,3-difluorobenzeneboronic acid, 4-Ethoxy-2,3-difluorophenylboronic acid, (4-Ethoxy-2,3-difluorophenyl)boronic acid
PC7277-1G
APOSPC7277-1GEA
176
EUR
APOSPC7277-1G
2,3-Difluor-4-ethoxyphenylboronsäure 98%
2,3-Difluor-4-ethoxyphenylboronsäure
Formel:
C₈H₉BF₂O₃ Molecular Weight: 201,96 g/mol Schmelzpunkt: 151…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258743 CAS-Nummer: 212386-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...