3,4,5-Trifluorbenzoylchlorid 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3,4,5-Trifluorbenzoesäurechlorid

PC7285K-1G
APOSPC7285K-1GEA 21.9 EUR
APOSPC7285K-1G
3,4,5-Trifluorbenzoylchlorid 97%
3,4,5-Trifluorbenzoylchlorid
Formel: C₇H₂ClF₃O
Molecular Weight: 194,54 g/mol
Siedepunkt: 173…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00083526
CAS-Nummer: 177787-26-7
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR