1,1'-Carbonylbis(3-methyl-1H-imidazol-3-ium) bis(trifluoromethanesulphonate)

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,1'-Carbonylbis(3-methyl-1H-imidazol-3-ium) ditriflate

PC7301-250MG
APOSPC7301-250MGEA 377 EUR
APOSPC7301-250MG
1,1'-Carbonylbis(3-methyl-1H-imidazol-3-ium) bis(trifluoromethanesulphonate)
1,1'-Carbonylbis(3-methyl-1H-imidazol-3-ium) bis(trifluoromethanesulphonate)

An efficient reagent for aminoacylations.

Formel: C₉H₁₂N₄O·2CF₃O₃S
Molecular Weight: 490,356 g/mol
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00269944
CAS-Nummer: 120418-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR