4-Deoxy-4-fluoro-D-glucose

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-4-deoxy-d-glucopyranose

PC7308-25MG
APOSPC7308-25MGEA 242 EUR
APOSPC7308-25MG
4-Deoxy-4-fluoro-D-glucose
4-Deoxy-4-fluoro-D-glucose
Formel: C₆H₁₁FO₅
Molecular Weight: 182,147 g/mol
Schmelzpunkt: 189…190 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00077529
CAS-Nummer: 29218-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR