2,5-Bis(trifluormethyl)benzonitril
Lieferant: Apollo Scientific
|
Gefahr
|
PC7321-1G
PC7321-25G
PC7321-5G
APOSPC7321-1GEA
52.2
EUR
APOSPC7321-1G
APOSPC7321-25G
APOSPC7321-5G
2,5-Bis(trifluormethyl)benzonitril
2,5-Bis(trifluormethyl)benzonitril
Formel:
C₉H₃F₆N Siedepunkt: 194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094423 CAS-Nummer: 51012-27-2 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...