1-[4-Nitro-3-(trifluoromethyl)phenyl]homopiperazine hydrochloride

Lieferant: Apollo Scientific

PC7328-500MG
APOSPC7328-500MGEA 400 EUR
APOSPC7328-500MG
1-[4-Nitro-3-(trifluoromethyl)phenyl]homopiperazine hydrochloride
1-[4-Nitro-3-(trifluoromethyl)phenyl]homopiperazine hydrochloride
Formel: C₁₂H₁₅ClF₃N₃O₂
Schmelzpunkt: 216…219 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025926
CAS-Nummer: 1185298-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR