1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine-3-carboxylic acid

Lieferant: Apollo Scientific

PC7331-1G
APOSPC7331-1GEA 0 EUR
APOSPC7331-1G
1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine-3-carboxylic acid
1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine-3-carboxylic acid
Formel: C₁₃H₁₃F₃N₂O₄
Schmelzpunkt: 138…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01831167
CAS-Nummer: 325702-09-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR