1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylic acid

Lieferant: Apollo Scientific

PC7332-1G
APOSPC7332-1GEA 286 EUR
APOSPC7332-1G
1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylic acid
1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylic acid
Formel: C₁₃H₁₃F₃N₂O₄
Schmelzpunkt: 137…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01908499
CAS-Nummer: 325702-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR