8-[5-(Trifluoromethyl)pyridin-2-yl]-1,4-dioxa-8-azaspiro[4.5]decane

Lieferant: Apollo Scientific

PC7334-1G
APOSPC7334-1GEA 257 EUR
APOSPC7334-1G
8-[5-(Trifluoromethyl)pyridin-2-yl]-1,4-dioxa-8-azaspiro[4.5]decane
8-[5-(Trifluoromethyl)pyridin-2-yl]-1,4-dioxa-8-azaspiro[4.5]decane
Formel: C₁₃H₁₅F₃N₂O₂
Schmelzpunkt: 95…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692463
CAS-Nummer: 942474-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR