2-[4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl]ethanol

Lieferant: Apollo Scientific

Synonyme: 1-(2-Hydroxyethyl)-4-[5-(trifluoromethyl)pyridin-2-yl]piperazine

PC7336-1G PC7336-10G PC7336-5G
APOSPC7336-1GEA 125 EUR
APOSPC7336-1G APOSPC7336-10G APOSPC7336-5G
2-[4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl]ethanol
2-[4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl]ethanol
Formel: C₁₂H₁₆F₃N₃O
Molecular Weight: 275.28 g/mol
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00203918
CAS-Nummer: 215434-37-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR