1-[5-(Trifluoromethyl)pyridin-2-yl]piperidine-3-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 2-(3-Carboxypiperidin-1-yl)-5-(trifluoromethyl)pyridine, 3-Carboxy-1-[5-(trifluoromethyl)pyridin-2-yl]piperidine

PC7337-1G
APOSPC7337-1GEA 236 EUR
APOSPC7337-1G
1-[5-(Trifluoromethyl)pyridin-2-yl]piperidine-3-carboxylic acid
1-[5-(Trifluoromethyl)pyridin-2-yl]piperidine-3-carboxylic acid
Formel: C₁₂H₁₃F₃N₂O₂
Molecular Weight: 274,239 g/mol
Schmelzpunkt: 99…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02091544
CAS-Nummer: 327971-41-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR