1-[5-(Trifluoromethyl)pyridin-2-yl]piperidine-4-carboxamide
Lieferant: Apollo Scientific
PC7344-1G
APOSPC7344-1GEA
303
EUR
APOSPC7344-1G
1-[5-(Trifluoromethyl)pyridin-2-yl]piperidine-4-carboxamide
1-[5-(Trifluoromethyl)pyridin-2-yl]piperidine-4-carboxamide
Formel:
C₁₂H₁₄F₃N₃O Schmelzpunkt: 190…192 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00205858 CAS-Nummer: 792940-04-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...