2,4-Dimethoxy-5-fluoroaniline
Lieferant: Apollo Scientific
|
Gefahr
|
PC7349-1G
PC7349-250MG
PC7349-5G
APOSPC7349-1GEA
0
EUR
APOSPC7349-1G
APOSPC7349-250MG
APOSPC7349-5G
2,4-Dimethoxy-5-fluoroaniline
2,4-Dimethoxy-5-fluoroaniline
Formel:
C₈H₁₀FNO₂ Schmelzpunkt: 47…49 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12547861 CAS-Nummer: 195136-65-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...