7-Fluoro-1H-indazole-3-carboxylic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC7358-250MG
APOSPC7358-250MGEA
102
EUR
APOSPC7358-250MG
7-Fluoro-1H-indazole-3-carboxylic acid 95+%
7-Fluoro-1H-indazole-3-carboxylic acid
Formel:
C₈H₅FN₂O₂ Molecular Weight: 180,14 g/mol Siedepunkt: 445 °C Dichte: 1,611 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD07378920 CAS-Nummer: 959236-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...