Methyl-7-fluoro-1H-indazole-3-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 7-fluoro-1H-indazole-3-carboxylate

PC7359-250MG
APOSPC7359-250MGEA 116 EUR
APOSPC7359-250MG
Methyl-7-fluoro-1H-indazole-3-carboxylate 95%
Methyl-7-fluoro-1H-indazole-3-carboxylate
Formel: C₉H₇FN₂O₂
Molecular Weight: 194,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16170462
CAS-Nummer: 932041-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR