4'-Brom-3'-fluoracetanilid
Lieferant: Apollo Scientific
|
Achtung
|
PC7364-25G
PC7364-5G
PC7364-1G
APOSPC7364-25GEA
238
EUR
APOSPC7364-25G
APOSPC7364-5G
APOSPC7364-1G
4'-Brom-3'-fluoracetanilid
4'-Brom-3'-fluoracetanilid
Formel:
H₃CC(O)NH(C₆H₃BrF) Schmelzpunkt: 149…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672937 CAS-Nummer: 351-30-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...