2-(2-Bromo-4-fluorophenoxy)propane 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-4-fluoro-1-isopropoxybenzene
PC7408-1G
APOSPC7408-1GEA
111
EUR
APOSPC7408-1G
2-(2-Bromo-4-fluorophenoxy)propane 98%
2-(2-Bromo-4-fluorophenoxy)propane
Formel:
C₉H₁₀BrFO Molecular Weight: 233,078 g/mol Siedepunkt: 243 °C (760 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070761 CAS-Nummer: 202865-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...