3'-Brom-4'-fluorpropiophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC7411-1G
APOSPC7411-1GEA
92.8
EUR
APOSPC7411-1G
3'-Brom-4'-fluorpropiophenon 98%
3'-Brom-4'-fluorpropiophenon
Formel:
C₉H₈BrFO Schmelzpunkt: 60…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143012 CAS-Nummer: 202865-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...