3-(Trifluormethoxy)benzamid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC7435-5G
PC7435-1G
APOSPC7435-5GEA
0
EUR
APOSPC7435-5G
APOSPC7435-1G
3-(Trifluormethoxy)benzamid 98%
3-(Trifluormethoxy)benzamid
Formel:
C₈H₆F₃NO₂ Schmelzpunkt: 115…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041512 CAS-Nummer: 658-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...