4-(Trifluormethoxy)benzamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Trifluormethoxy)benzoesäureamid

PC7436-1G
APOSPC7436-1GEA 21.9 EUR
APOSPC7436-1G
4-(Trifluormethoxy)benzamid 97%
4-(Trifluormethoxy)benzamid
Formel: C₈H₆F₃NO₂
Molecular Weight: 205,14 g/mol
Schmelzpunkt: 155…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041384
CAS-Nummer: 456-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR