4-(Trifluormethoxy)benzamidoxim 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC7436D-25G
PC7436D-5G
PC7436D-1G
APOSPC7436D-25GEA
0
EUR
APOSPC7436D-25G
APOSPC7436D-5G
APOSPC7436D-1G
4-(Trifluormethoxy)benzamidoxim 97%
4-(Trifluormethoxy)benzamidoxim
Formel:
C₈H₇F₃N₂O₂ Schmelzpunkt: 113…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085175 CAS-Nummer: 56935-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...