4-(Trifluormethoxy)benzoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC7438-100G
PC7438-25G
APOSPC7438-100GEA
0
EUR
APOSPC7438-100G
APOSPC7438-25G
4-(Trifluormethoxy)benzoesäure 97%
4-(Trifluormethoxy)benzoesäure
Formel:
C₈H₅F₃O₃ Schmelzpunkt: 150…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002541 CAS-Nummer: 330-12-1 EINECS: 206-352-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...