3-(Trifluormethoxy)benzoylchlorid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC7438M-5G
APOSPC7438M-5GEA
94.4
EUR
APOSPC7438M-5G
3-(Trifluormethoxy)benzoylchlorid 97%
3-(Trifluormethoxy)benzoylchlorid
Formel:
C₈H₄ClF₃O₂ Molecular Weight: 224.57 g/mol Dichte: 1.43 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061268 CAS-Nummer: 86270-03-3 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...