3-(Trifluormethoxy)benzylamin 97%

Lieferant: Apollo Scientific
Gefahr

PC7438O-1G PC7438O-25G PC7438O-5G
APOSPC7438O-1GEA 0 EUR
APOSPC7438O-1G APOSPC7438O-25G APOSPC7438O-5G
3-(Trifluormethoxy)benzylamin 97%
3-(Trifluormethoxy)benzylamin
Formel: C₈H₈F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00061267
CAS-Nummer: 93071-75-1
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR