4-(Trifluoromethoxy)phenoxyacetic hydrazide 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC7439PC-1G
PC7439PC-250MG
APOSPC7439PC-1GEA
0
EUR
APOSPC7439PC-1G
APOSPC7439PC-250MG
4-(Trifluoromethoxy)phenoxyacetic hydrazide 97%
4-(Trifluoromethoxy)phenoxyacetic hydrazide
Formel:
C₉H₉F₃N₂O₃ Schmelzpunkt: 133…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052355 CAS-Nummer: 175204-36-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...