4-(Trifluormethoxy)phenylhydrazin Hydrochlorid 97%

Lieferant: Apollo Scientific
Achtung

PC7439PT-100G PC7439PT-25G PC7439PT-5G
APOSPC7439PT-100GEA 0 EUR
APOSPC7439PT-100G APOSPC7439PT-25G APOSPC7439PT-5G
4-(Trifluormethoxy)phenylhydrazin Hydrochlorid 97%
4-(Trifluormethoxy)phenylhydrazin Hydrochlorid
Formel: C₇H₈ClF₃N₂O
Schmelzpunkt: 230 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00053033
CAS-Nummer: 133115-72-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR