2-(Trifluoromethoxy)terephthalic acid 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
PC7439TK-1G
APOSPC7439TK-1GEA
0
EUR
APOSPC7439TK-1G
2-(Trifluoromethoxy)terephthalic acid 95+%
2-(Trifluoromethoxy)terephthalic acid
Formel:
C₉H₅F₃O₅ Schmelzpunkt: 270…272 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00203986 CAS-Nummer: 175278-21-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...