4-(Trifluormethoxy)thiobenzamid
Lieferant: Apollo Scientific
|
Achtung
|
PC7439U-10G
PC7439U-5G
APOSPC7439U-10GEA
400
EUR
APOSPC7439U-10G
APOSPC7439U-5G
4-(Trifluormethoxy)thiobenzamid
4-(Trifluormethoxy)thiobenzamid
Formel:
C₈H₆F₃NOS Schmelzpunkt: 124…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068113 CAS-Nummer: 149169-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...